The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026888.3 | NP_081164.2 | 7 - 8 | 775 | 63 |
GenBank mRNA | AA198694.1 | - | 2 - 3 | 189 | 63 |
AK005277.1 | - | 7 - 8 | 720 | 63 | |
BC138913.1 | - | 7 - 8 | 689 | 63 | |
BC145734.1 | - | 7 - 8 | 689 | 63 |