The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183147.2 | NP_898970.1 | 1 - 2 | 108 | 65 |
GenBank mRNA | AK049995.1 | - | 1 - 2 | 145 | 65 |
BC056484.1 | - | 1 - 2 | 108 | 65 |