Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ipo7
Entrez Gene ID
233726
Gene Name
importin 7
Gene Aliases
A330055O14Rik, C330016G14, Imp7, Ranbp7
Chromosome Location
Chr.7: 110018227 - 110056610 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181517.3 NP_852658.2 2 - 3 168 110
GenBank mRNA AK039534.1 - 2 - 3 320 110
AK053498.1 - 2 - 3 315 110
AK079903.1 - 2 - 3 279 110
AK148820.1 - 2 - 3 110
AY316153.1 - 2 - 3 168 110

View Full Product Details