Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a7
Entrez Gene ID
63993
Gene Name
solute carrier family 5 (choline transporter), member 7
Gene Aliases
CHT1
Chromosome Location
Chr.17: 54273590 - 54299041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022025.4 NP_071308.2 4 - 5 958 80
GenBank mRNA AF276872.2 - 3 - 4 449 80
AJ401467.1 - 3 - 4 449 80
AK034415.1 - 4 - 5 842 80
AK053063.1 - 4 - 5 960 80
BC065089.1 - 3 - 4 475 80

View Full Product Details