The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028232.2 | NP_082508.1 | 1 - 2 | 101 | 122 |
GenBank mRNA | AK154188.1 | - | 1 - 2 | 120 | 122 |