Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ring1
Entrez Gene ID
19763
Gene Name
ring finger protein 1
Gene Aliases
Ring1A
Chromosome Location
Chr.17: 34020792 - 34024680 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009066.3 NP_033092.3 3 - 4 459 83
GenBank mRNA AK154962.1 - 3 - 4 420 83
AK155509.1 - 3 - 4 435 83
BC009070.1 - 3 - 4 422 83
BC082771.1 - 1 - 2 637 83
BU614216.1 - 3 - 4 379 83
CT010219.1 - 1 - 2 149 83
CV560731.1 - 1 - 2 92 83
KJ650216.1 - 2 - 3 236 83
Y12881.1 - 1 - 2 155 83

View Full Product Details