Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim31
Entrez Gene ID
224762
Gene Name
tripartite motif-containing 31
Gene Aliases
BC026666, HCG1, HCGI, RNF
Chromosome Location
Chr.17: 36898130 - 36910217 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146077.2 NP_666189.1 2 - 3 737 137
GenBank mRNA AK033650.1 - 2 - 3 750 137
BC026666.1 - 2 - 3 737 137

View Full Product Details