Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scly
Entrez Gene ID
50880
Gene Name
selenocysteine lyase
Gene Aliases
9830169H08, A930015N15Rik, SCL, Scly1, Scly2
Chromosome Location
Chr.1: 91298295 - 91321080 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016717.3 NP_057926.2 1 - 2 114 81
GenBank mRNA AF175407.1 - 1 - 2 66 81
AK036533.1 - 1 - 2 83 81
AK036750.1 - 1 - 2 116 81
BC019879.1 - 1 - 2 74 81
BC021389.1 - 1 - 2 98 81

View Full Product Details