The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136070.1 | NP_001129542.1 | 1 - 2 | 216 | 71 |
NM_010709.3 | NP_034839.2 | 1 - 2 | 216 | 71 | |
GenBank mRNA | AK030312.1 | - | 1 - 2 | 228 | 71 |
AK034115.1 | - | 1 - 2 | 188 | 71 | |
AK036154.1 | - | 1 - 2 | 212 | 71 | |
AK046539.1 | - | 1 - 2 | 228 | 71 | |
AK047005.1 | - | 1 - 2 | 211 | 71 | |
AK078472.1 | - | 1 - 2 | 214 | 71 | |
AK082748.1 | - | 1 - 2 | 231 | 71 | |
AK083638.1 | - | 1 - 2 | 176 | 71 | |
AK139052.1 | - | 1 - 2 | 216 | 71 | |
AK161269.1 | - | 1 - 2 | 199 | 71 | |
AK166085.1 | - | 1 - 2 | 228 | 71 | |
AK167980.1 | - | 1 - 2 | 212 | 71 | |
AK168120.1 | - | 1 - 2 | 182 | 71 | |
AK170252.1 | - | 1 - 2 | 243 | 71 | |
AK170448.1 | - | 1 - 2 | 228 | 71 | |
AK171115.1 | - | 1 - 2 | 194 | 71 | |
BC023709.1 | - | 1 - 2 | 222 | 71 | |
BC025036.1 | - | 1 - 2 | 172 | 71 | |
U58337.1 | - | 1 - 2 | 101 | 71 |