Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Elf2
- Entrez Gene ID
-
69257
- Gene Name
- E74-like factor 2
- Gene Aliases
-
2610036A20Rik, A230104O07Rik, AW111824, BB183398, EU32, NERF, NERF-1A, NERF-1B, NERF-2
- Chromosome Location
-
Chr.3: 51252720 - 51340644 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC051937.1 |
- |
3 - 4 |
486 |
76 |
View Full Product Details