Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrriq4
Entrez Gene ID
68307
Gene Name
leucine-rich repeats and IQ motif containing 4
Gene Aliases
4930558O21Rik
Chromosome Location
Chr.3: 30644506 - 30672431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290510.1 NP_001277439.1 1 - 2 373 65
GenBank mRNA AK015024.1 - 1 - 2 324 65

View Full Product Details