The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290510.1 | NP_001277439.1 | 5 - 6 | 1931 | 75 |
NM_026668.2 | NP_080944.2 | 7 - 8 | 2094 | 75 | |
GenBank mRNA | AK015024.1 | - | 5 - 6 | 1882 | 75 |
AK015586.1 | - | 1 - 2 | 106 | 75 | |
AK019775.1 | - | 7 - 8 | 2095 | 75 | |
BC145806.1 | - | 7 - 8 | 1795 | 75 |