Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spata16
Entrez Gene ID
70862
Gene Name
spermatogenesis associated 16
Gene Aliases
4921511F01Rik, 4930503K02Rik, Nyd-sp12
Chromosome Location
Chr.3: 26637631 - 26983212 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027583.3 NP_081859.1 5 - 6 1057 91
NM_029150.4 NP_083426.1 5 - 6 1057 91
GenBank mRNA AK014869.1 - 5 - 6 1021 91
AK076628.1 - 5 - 6 1058 91
BC100443.1 - 5 - 6 1038 91
BC132205.1 - 5 - 6 981 91
BC132539.1 - 5 - 6 981 91

View Full Product Details