Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nceh1
- Entrez Gene ID
-
320024
- Gene Name
- neutral cholesterol ester hydrolase 1
- Gene Aliases
-
Aadacl1, B230106I24Rik, CPO-BP, Nceh
- Chromosome Location
-
Chr.3: 27183004 - 27245266 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK038691.1 |
- |
- |
516 |
69 |
View Full Product Details