Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpinb10
Entrez Gene ID
241197
Gene Name
serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10
Gene Aliases
9830131G07, BB233602, Serpinb10-ps
Chromosome Location
Chr.1: 107528994 - 107560735 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160307.1 NP_001153779.1 5 - 6 556 107
NM_198028.3 NP_932145.3 6 - 7 732 107
GenBank mRNA AK036537.1 - 6 - 7 710 107
AK052580.1 - 5 - 6 557 107

View Full Product Details