Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tdrd5
Entrez Gene ID
214575
Gene Name
tudor domain containing 5
Gene Aliases
Gm103
Chromosome Location
Chr.1: 156255296 - 156305295 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134741.1 NP_001128213.1 10 - 11 1804 80
NM_001277730.1 NP_001264659.1 10 - 11 1510 80
GenBank mRNA AK132674.1 - 10 - 11 1790 80
AK133026.1 - 8 - 9 1175 80
AY522557.1 - 4 - 5 664 80
BC099972.1 - 10 - 11 1510 80

View Full Product Details