Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntnap1
Entrez Gene ID
53321
Gene Name
contactin associated protein-like 1
Gene Aliases
AI841080, Caspr, NCP1, Nrxn4, p190, shm
Chromosome Location
Chr.11: 101174629 - 101190727 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016782.2 NP_058062.2 1 - 2 209 82
GenBank mRNA AF039833.1 - 1 - 2 219 82

View Full Product Details