The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008911.2 | NP_032937.1 | 5 - 6 | 794 | 55 |
GenBank mRNA | AK142632.1 | - | 5 - 6 | 746 | 55 |