The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028148.2 | NP_082424.2 | 2 - 3 | 325 | 66 |
GenBank mRNA | AK129478.1 | - | 2 - 3 | 217 | 66 |
AK163564.1 | - | 2 - 3 | 187 | 66 | |
BC058682.1 | - | 2 - 3 | 306 | 66 |