The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079849.2 | NP_001073318.1 | 2 - 3 | 214 | 75 |
NM_145457.4 | NP_663432.1 | 2 - 3 | 432 | 75 | |
GenBank mRNA | AK040012.1 | - | 2 - 3 | 216 | 75 |
BC019726.1 | - | 2 - 3 | 403 | 75 | |
BC051042.1 | - | 2 - 3 | 389 | 75 | |
CB840007.1 | - | 1 - 2 | 187 | 75 | |
CD363907.1 | - | 2 - 3 | 190 | 75 | |
CJ173734.1 | - | 2 - 3 | 418 | 75 |