Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Samd14
Entrez Gene ID
217125
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
AI839049, AI854782, BC034054
Chromosome Location
Chr.11: 95009402 - 95026087 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146025.2 NP_666137.1 5 - 6 1300 91
GenBank mRNA AK044765.1 - 5 - 6 829 91
AK047184.1 - 5 - 6 1277 91
AK049753.1 - 5 - 6 1303 91
BC034054.1 - 5 - 6 824 91
BC049954.1 - 5 - 6 857 91

View Full Product Details