Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rnps1
- Entrez Gene ID
-
19826
- Gene Name
- ribonucleic acid binding protein S1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 24414675 - 24425898 on Build GRCm38
- Gene Symbol
- Evpl
- Entrez Gene ID
-
14027
- Gene Name
- envoplakin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 116220559 - 116238147 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC014864.1 |
- |
- |
|
108 |
View Full Product Details