The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289898.1 | NP_001276827.1 | 2 - 3 | 220 | 73 |
NM_001289899.1 | NP_001276828.1 | 2 - 3 | 692 | 73 | |
NM_013767.6 | NP_038795.3 | 2 - 3 | 224 | 73 | |
GenBank mRNA | AB028241.1 | - | 2 - 3 | 73 | |
AK158709.1 | - | 2 - 3 | 693 | 73 | |
BC026127.1 | - | 2 - 3 | 173 | 73 |