Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Metrn
Entrez Gene ID
70083
Gene Name
meteorin, glial cell differentiation regulator
Gene Aliases
1810034B16Rik, Hyrac
Chromosome Location
Chr.17: 25794571 - 25797094 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133719.2 NP_598480.1 1 - 2 184 104
GenBank mRNA AK007688.1 - 1 - 2 124 104
AK087402.1 - 1 - 2 185 104
BC008695.1 - 1 - 2 62 104
BC037181.1 - 1 - 2 111 104
BC132177.1 - 2 - 3 186 104
DQ133462.1 - 1 - 2 107 104

View Full Product Details