Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smoc1
Entrez Gene ID
64075
Gene Name
SPARC related modular calcium binding 1
Gene Aliases
2600002F22Rik, AI848508, SRG
Chromosome Location
Chr.12: 81026808 - 81186414 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146217.1 NP_001139689.1 9 - 10 1225 94
NM_022316.2 NP_071711.2 9 - 10 1192 94
GenBank mRNA AF070470.1 - 9 - 10 1168 94
AK040931.1 - 9 - 10 1226 94
BC031804.1 - 9 - 10 1172 94

View Full Product Details