Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mfsd5
Entrez Gene ID
106073
Gene Name
major facilitator superfamily domain containing 5
Gene Aliases
AA409375, AW556797, D15Mgi27
Chromosome Location
Chr.15: 102279456 - 102281744 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC009140.1 - 1 - 1 31 63

View Full Product Details