Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sept3
Entrez Gene ID
24050
Gene Name
septin 3
Gene Aliases
3110018K01Rik, AV154067, B530002E20Rik, Sep3
Chromosome Location
Chr.15: 82272451 - 82294574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK046722.1 - 10 - 11 1213 58

View Full Product Details