Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eml2
Entrez Gene ID
72205
Gene Name
echinoderm microtubule associated protein like 2
Gene Aliases
1600029N02Rik
Chromosome Location
Chr.7: 19176266 - 19206482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162996.1 NP_001156468.1 12 - 13 1290 90
NM_028153.1 NP_082429.1 12 - 13 1347 90
GenBank mRNA AK005562.1 - 12 - 13 1313 90
BC055476.1 - 12 - 13 1290 90

View Full Product Details