The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176971.2 | NP_795945.1 | 2 - 3 | 212 | 79 |
GenBank mRNA | AK034442.1 | - | 2 - 3 | 213 | 79 |
AK049693.1 | - | 2 - 3 | 239 | 79 | |
AK161867.1 | - | 2 - 3 | 429 | 79 | |
BC107383.2 | - | 2 - 3 | 184 | 79 |