Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lalba
Entrez Gene ID
16770
Gene Name
lactalbumin, alpha
Gene Aliases
AW208827
Chromosome Location
Chr.15: 98480400 - 98482934 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010679.1 NP_034809.1 2 - 3 309 70
GenBank mRNA BC006922.1 - 2 - 3 321 70
BC069916.1 - 2 - 3 309 70
M80909.1 - 2 - 3 309 70

View Full Product Details