The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171034.1 | NP_001164505.1 | 9 - 10 | 950 | 76 |
NM_001171035.1 | NP_001164506.1 | 8 - 9 | 806 | 76 | |
NM_001171036.1 | NP_001164507.1 | 9 - 10 | 771 | 76 | |
NM_026669.4 | NP_080945.1 | 8 - 9 | 735 | 76 | |
GenBank mRNA | AK019865.2 | - | 8 - 9 | 697 | 76 |
AK036782.1 | - | 8 - 9 | 697 | 76 | |
AK036994.1 | - | 8 - 9 | 697 | 76 | |
AK049604.1 | - | 8 - 9 | 686 | 76 | |
AK050299.1 | - | 8 - 9 | 698 | 76 | |
AK050323.1 | - | 8 - 9 | 686 | 76 | |
AK088030.1 | - | 8 - 9 | 697 | 76 | |
AK088683.1 | - | 8 - 9 | 698 | 76 | |
AK139901.1 | - | 8 - 9 | 698 | 76 | |
AK159380.1 | - | 8 - 9 | 697 | 76 | |
AK166935.1 | - | 8 - 9 | 697 | 76 | |
AK206236.1 | - | 3 - 4 | 202 | 76 | |
BC005588.1 | - | 8 - 9 | 680 | 76 |