Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ipo9
Entrez Gene ID
226432
Gene Name
importin 9
Gene Aliases
0710008K06Rik, AI845704, C78347, Imp9, Ranbp9, mKIAA1192
Chromosome Location
Chr.1: 135382315 - 135430502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153774.1 NP_722469.1 22 - 23 2995 76
GenBank mRNA AF273673.1 - 22 - 23 2943 76
AJ309238.1 - 22 - 23 2937 76
AK036687.1 - 22 - 23 2945 76
BC056469.1 - 23 - 24 3143 76
BC079546.1 - 24 - 25 3110 76
BC098508.1 - 22 - 23 2945 76

View Full Product Details