Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd13c
Entrez Gene ID
433667
Gene Name
ankyrin repeat domain 13c
Gene Aliases
AI505652, AU022220
Chromosome Location
Chr.3: 157946668 - 158006837 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013806.1 NP_001013828.1 12 - 13 1559 81
GenBank mRNA AK135906.1 - 13 - 14 1295 81
BC065088.1 - 11 - 12 3753 81
BC138991.1 - 12 - 13 1567 81
BC138992.1 - 13 - 14 1546 81
CF736962.1 - 5 - 6 555 81

View Full Product Details