Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ap3b2
Entrez Gene ID
11775
Gene Name
adaptor-related protein complex 3, beta 2 subunit
Gene Aliases
AI549966, AU042881, Naptb, [b]-NAP, beta-NAP, beta3B
Chromosome Location
Chr.7: 81460399 - 81494038 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021492.3 NP_067467.2 24 - 25 3264 90
GenBank mRNA AB030202.1 - 10 - 11 1138 90
AY528675.1 - 24 - 25 3045 90
BC030306.1 - 7 - 8 928 90
BC139378.1 - 24 - 25 3012 90
BC139379.1 - 24 - 25 3012 90

View Full Product Details