The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016697.3 | NP_057906.2 | 7 - 8 | 1741 | 71 |
GenBank mRNA | AF185614.1 | - | 7 - 8 | 1681 | 71 |
AK146702.1 | - | 7 - 8 | 1718 | 71 | |
AK159394.1 | - | 7 - 8 | 1710 | 71 | |
AK159435.1 | - | 7 - 8 | 1710 | 71 | |
AK159764.1 | - | 7 - 8 | 1718 | 71 | |
AK168402.1 | - | 7 - 8 | 1719 | 71 | |
BC036126.1 | - | 7 - 8 | 1688 | 71 |