Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Enah
Entrez Gene ID
13800
Gene Name
enabled homolog (Drosophila)
Gene Aliases
Mena, NDPP-1, Ndpp1, WBP8
Chromosome Location
Chr.1: 181896384 - 182019980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083120.2 NP_001076589.1 7 - 8 2311 82
NM_001083121.2 NP_001076590.1 7 - 8 1579 82
NM_008680.3 NP_032706.2 8 - 9 2323 82
NM_010135.3 NP_034265.2 8 - 9 2368 82
GenBank mRNA BC062927.1 - 7 - 8 1198 82
U72520.1 - 7 - 8 1272 82
U72521.1 - 7 - 8 1998 82
U72522.1 - 8 - 9 2010 82
U72523.1 - 8 - 9 2055 82

View Full Product Details