Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rps6kc1
Entrez Gene ID
320119
Gene Name
ribosomal protein S6 kinase polypeptide 1
Gene Aliases
AA682037, B130003F20Rik, C80612, Rpk118
Chromosome Location
Chr.1: 190772879 - 190912139 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178775.4 NP_848890.3 5 - 6 585 70
GenBank mRNA AK030313.1 - - 563 70
AK149858.1 - 5 - 6 550 70

View Full Product Details