Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubxn2a
Entrez Gene ID
217379
Gene Name
UBX domain protein 2A
Gene Aliases
6330407P03Rik, Ubxd4
Chromosome Location
Chr.12: 4876647 - 4907781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145441.3 NP_663416.1 3 - 4 365 121
GenBank mRNA AK031821.1 - 3 - 4 366 121
AK041556.1 - 3 - 4 402 121
AK134666.1 - 3 - 4 398 121
BC004632.1 - 3 - 4 317 121

View Full Product Details