The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK044741.1 | - | 1 - 2 | 216 | 73 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | G patch domain containing 2 | Gpatch2 | 5830433G22Rik, 5830436K05Rik, AI427714, AI447508, AW107440, AW491060, Gpatc2 | -78.4kb | Sense | Chr. - 187215393 - 187369480 on Build GRCm38 |
Downstream | predicted gene, 36388 | Gm36388 | 73.2kb | Anti-sense | Chr. - 187521696 - 187652285 on Build GRCm38 |