Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hhipl2
Entrez Gene ID
78772
Gene Name
hedgehog interacting protein-like 2
Gene Aliases
4930507C10Rik, AW208802, Hhip3
Chromosome Location
Chr.1: 183413929 - 183437053 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK019671.1 - 4 - 5 1281 67
BC034362.1 - 3 - 4 1026 67

View Full Product Details