Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spta1
Entrez Gene ID
20739
Gene Name
spectrin alpha, erythrocytic 1
Gene Aliases
AF093576, AI451697, Spna-1, Spna1, ha, ihj, nmf4, sph
Chromosome Location
Chr.1: 174172776 - 174248449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011465.4 NP_035595.2 46 - 47 6697 83
GenBank mRNA AF093576.1 - 46 - 47 6605 83
AK086017.1 - 3 - 4 337 83
AK143823.1 - 10 - 11 1365 83
AK161449.1 - 16 - 17 3623 83
BC150747.1 - 46 - 47 6562 83
U87455.1 - 46 - 47 6677 83

View Full Product Details