Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gipr
Entrez Gene ID
381853
Gene Name
gastric inhibitory polypeptide receptor
Gene Aliases
GIP-R, Gm1081, Gm160
Chromosome Location
Chr.7: 19156063 - 19170248 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080815.1 NP_001074284.1 11 - 12 1170 94
GenBank mRNA AK133386.1 - 5 - 6 1024 94
AK146552.1 - 9 - 10 1502 94

View Full Product Details