The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026578.3 | NP_080854.1 | 2 - 3 | 538 | 118 |
GenBank mRNA | AK021256.1 | - | 2 - 3 | 299 | 118 |
AK075923.1 | - | 1 - 2 | 273 | 118 | |
AK167001.1 | - | 2 - 3 | 278 | 118 | |
BC021873.1 | - | 3 - 4 | 402 | 118 | |
BC048685.1 | - | 2 - 3 | 457 | 118 |