Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rrh
Entrez Gene ID
20132
Gene Name
retinal pigment epithelium derived rhodopsin homolog
Gene Aliases
-
Chromosome Location
Chr.3: 129808575 - 129822505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK021379.1 - 1 - 2 128 60

View Full Product Details