Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctsc
Entrez Gene ID
13032
Gene Name
cathepsin C
Gene Aliases
AI047818, DPP1, DPPI
Chromosome Location
Chr.7: 88278023 - 88310875 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK152524.1 - 3 - 4 462 91

View Full Product Details