Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lrriq3
- Entrez Gene ID
-
74435
- Gene Name
- leucine-rich repeats and IQ motif containing 3
- Gene Aliases
-
4930438B07Rik, 4930511J15Rik, 4933403H06Rik, BB020367, Lrrc44
- Chromosome Location
-
Chr.3: 155093434 - 155194278 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015333.1 |
- |
6 - 7 |
1138 |
72 |
View Full Product Details