The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026819.3 | NP_081095.2 | 8 - 9 | 1072 | 64 |
GenBank mRNA | AK003958.1 | - | 8 - 9 | 962 | 64 |
AK146317.1 | - | 8 - 9 | 1019 | 64 | |
AK159964.1 | - | 8 - 9 | 994 | 64 | |
AK168115.1 | - | 8 - 9 | 1019 | 64 | |
AK168147.1 | - | 8 - 9 | 1019 | 64 | |
BC003930.1 | - | 8 - 9 | 1003 | 64 | |
BC023825.1 | - | 7 - 8 | 2113 | 64 |