Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lysmd4
- Entrez Gene ID
-
75099
- Gene Name
- LysM, putative peptidoglycan-binding, domain containing 4
- Gene Aliases
-
4930506D23Rik
- Chromosome Location
-
Chr.7: 67222544 - 67228468 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC030310.1 |
- |
3 - 4 |
339 |
92 |
View Full Product Details