The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191001.1 | NP_001177930.1 | 1 - 2 | 130 | 70 |
NM_152815.2 | NP_690028.2 | 1 - 2 | 130 | 70 | |
NR_034030.1 | 1 - 2 | 130 | 70 | ||
GenBank mRNA | AK012554.1 | - | 1 - 2 | 63 | 70 |
AK044132.1 | - | 1 - 2 | 71 | 70 | |
AK156066.1 | - | 1 - 2 | 99 | 70 | |
AK171170.1 | - | 1 - 2 | 57 | 70 | |
BC030943.1 | - | 1 - 2 | 55 | 70 |