The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191001.1 | NP_001177930.1 | 2 - 3 | 266 | 113 |
NM_152815.2 | NP_690028.2 | 2 - 3 | 266 | 113 | |
NR_034030.1 | 2 - 3 | 266 | 113 | ||
GenBank mRNA | AK012554.1 | - | 2 - 3 | 199 | 113 |
AK044132.1 | - | 2 - 3 | 207 | 113 | |
AK156066.1 | - | 2 - 3 | 235 | 113 | |
AK171170.1 | - | 2 - 3 | 193 | 113 | |
BC058648.1 | - | 2 - 3 | 169 | 113 |